Merck
27749-U
domnick hunter® NITROX UHP Hydrogen Generator
Model 40H, output flow, 0 ‑ 250 cc/min
ログイン組織・契約価格を表示する
サイズを選択してください
1 EA
¥2,140,000
¥2,140,000
Check Cart for Availability
バルクの問い合わせ
サイズを選択してください
表示を変更する
1 EA
¥2,140,000
About This Item
UNSPSCコード:
41112500
NACRES:
SB.54
¥2,140,000
Check Cart for Availability
バルクの問い合わせ
特徴
output flow 0 ‑ 250 cc/min
メーカー/製品名
Domnick Hunter 40H
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
domnick hunter hydrogen generators are a safe and economic alternative to high pressure gas cylinders. These generators deliver a continuous stream of 99.99999% pure hydrogen, suitable for use with any laboratory instrument requiring a supply of ultra high purity hydrogen, and ideal for carrier gas and fuel gas applications.
domnick hunter hydrogen generators use a proprietary ion exchange membrane to produce ultra-high-purity hydrogen from laboratory grade deionized water, totally eliminating the need for caustic solution.
These generators are designed for total safety and highly reliable operation. Hydrogen is generated only on demand, at low pressure, and minimal volume is stored within the unit. The unique digital display gives system status and diagnostic capabilities not available on any other competitive hydrogen generator.
CE Approved.
法的情報
NITROX is a trademark of domnick hunter group PLC
domnick hunter is a registered trademark of Parker Hannifin
最新バージョンのいずれかを選択してください:
試験成績書(COA)