Merck
22657
Supelco
SS-174 PTFE被膜付きセプタム
diam. 12.5 mm ( 1/2 in.)
ログイン組織・契約価格を表示する
サイズを選択してください
100 EA
¥29,400
¥29,400
Check Cart for Availability
バルクの問い合わせ
サイズを選択してください
表示を変更する
100 EA
¥29,400
About This Item
UNSPSCコード:
41115718
¥29,400
Check Cart for Availability
バルクの問い合わせ
物質
off-white silicone body (123 mil thick)
yellow PTFE face (2 mil thick)
包装
pkg of 100 ea
直径
12.5 mm ( 1/2 in.)
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
PTFE被膜付きセプタムは、より高度な不活性を必要とする用途向けに設計されています。オフホワイト色のシリコーンゴム本体(厚さ123 ミル)が、黄色のPTFE被膜(厚さ2ミル)を十分に支持しています。注入によってPTFEの表面が裂けると、いくらかのブリードが発生します。このブリードはゴムの露出部分が多いほど増えるため、ニードルガイドによって破裂部分の面積を減らすことをお勧めします。200°C~300°Cの入口温度で使用します。
PTFE-faced septa are suitable to overcome the degradation due to prolonged high temperatures which occurs in silicone rubber septa. The degradation results in baseline drift and additional peaks. It is generally fixed with the PTFE faced towards the column packing.[1]
最新バージョンのいずれかを選択してください:
試験成績書(COA)
文書ライブラリにアクセスする
Beatriz Cancho et al.
Journal of chromatography. A, 943(1), 1-13 (2002-02-01)
A headspace solid-phase microextraction (HS-SPME) procedure followed by gas chromatography and electron capture detection (GC-ECD) has been developed for the determination of aldehydes in drinking water samples at microg/l concentrations. A previous derivatization with o-(2,3,4,5,6-pentafluorobenzyl)hydroxylamine hydrochloride (PFBHA) was performed due
Carmen López-Blanco et al.
Analytical and bioanalytical chemistry, 383(4), 557-561 (2005-09-01)
Water contamination due to the wide variety of pesticides used in agriculture practices is a global environmental pollution problem. Analytical methods with low quantification limits are necessary. The application of a new extraction technique, solvent drop microextraction (SDME), followed by
Ian A. Fowlis
Gas Chromatography, 62-62 (2008)