Merck
Z743544
Microscope slide wells, 2-wells
Microscope slide wells, 2-wells
1のうち1の評価者がサンプル商品を受け取ったか、プロモーションに参加しました
well diam. × depth 1 cm × 0.50 mm, pack of 25
ログイン組織・契約価格を表示する
サイズを選択してください
25 EA
¥13,090
¥13,090
Check Cart for Availability
サイズを選択してください
表示を変更する
25 EA
¥13,090
About This Item
UNSPSCコード:
41121800
NACRES:
MA.01
¥13,090
Check Cart for Availability
包装
pack of 25
メーカー/製品名
Diversified Biotech MIWE-2000
ウェル直径×深さ
1 cm × 0.50 mm
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Pre-cut, adhesive well stickers that firmly adhere to microscope slides. Examine specimens with or without cover slips.
特徴および利点
  • Convenient - Pre-cut, firmly adhere to microscope slides and enable specimen examination with or without cover slips.
  • Multiple Applications - Examine or count tissue culture cells, yeast, bacteria and other microorganisms.
  • Versatile - Compatible with different styles of microscopes (conventional, inverted or dissecting).
  • Chemically Inert - Made of polyester with an acrylic adhesive. Microscope Slide-Wells are inert and stable under most laboratory conditions.
最新バージョンのいずれかを選択してください:
試験成績書(COA)
文書ライブラリにアクセスする
質問
レビュー
1のうち1の評価者がサンプル商品を受け取ったか、プロモーションに参加しました
アクティブなフィルタ
    • 1件のレビュー
    • 0件の投票
    星1/5個です。
    Supply chain coordinator
    These slides did not come with 2 wells but you have to attach an adhesive 2-well attachment to each slide which is very time consuming and not helpful when you use MANY slides for patient testing. Description says nothing about the 2-wells being a separate adhesive strip needing to be manually added to the slide.
    Googleで翻訳する
    役に立ちましたか?
    1. Sigma-Aldrich Corporationからの回答:
      Thank you for taking the time to leave a review! We are sorry to hear that your experience did meet expectations. We have provided this information to our internal product teams so that we can update our product information. We also encourage customers who experience problems to contact us by visiting https://www.sigmaaldrich.com/support/customer-support and submit a Report Product Issues ticket. We apologize for any inconvenience and look forward to hearing from you soon!