Merck
Z740379
Eco-Friendly SpecTainer, tamper evident, biodegradable
size 120 mL, sterile
ログイン組織・契約価格を表示する
サイズを選択してください
300 EA
¥39,280
¥39,280
Check Cart for Availability
サイズを選択してください
表示を変更する
300 EA
¥39,280
About This Item
UNSPSCコード:
41121800
NACRES:
NB.22
¥39,280
Check Cart for Availability
物質
polyethylene cap
polypropylene container
無菌性
sterile
包装
pack of 300 ea
メーカー/製品名
Simport C566-120AQSECO
サイズ
120 mL
類似した製品をお探しですか? 訪問 製品比較ガイド
関連するカテゴリー
詳細
The Simport Eco-Friendly SpecTainer will biodegrade to become some of the soil′s organic components in less than 7 years in constant contact with the soil instead of up to 400 years when using conventional plastics.
The Simport Eco-Friendly SpecTainer, one of the most innovative specimen containers on the market today, is now available in a new version: The Eco-Friendly SpecTainer, a rapidly degradable Specimen Container. Plastics take hundreds of years to degrade naturally in the environment. The only condition that is necessary for the Eco-Friendly SpecTainer to biodegrade is constant contact with other degrading material. No heat, physical stress, oxygen or sunlight necessary. The Eco-Friendly SpecTainer has the ability to break down safely and quickly, by biological means, into the raw materials of nature and disappear into the environment.
Does not contain heavy metals. No negative effect to its physical properties. No heat, light or mechanical stress needed for product breakdown. No toxic residue. Fully degradable within 7 years, aerobically or anaerobically.
法的情報
SpecTainer is a trademark of Simport
最新バージョンのいずれかを選択してください:
試験成績書(COA)