TMO383410-42
Nalgene® Certified Clean Biotainer(TM) Carboys
polycarbonate, volume 10 L, case of 2 ea
別名:
biotainer carboy, certified nalgene carboy, sterile carboy
ログイン組織・契約価格を表示する
サイズを選択してください
1 EACH
¥51,030
¥51,030
Check Cart for Availability
すべての画像(1)
サイズを選択してください
表示を変更する
1 EACH
¥51,030
About This Item
UNSPSCコード:
41121800
NACRES:
NB.43
¥51,030
Check Cart for Availability
おすすめの製品
物質
polycarbonate
包装
case of 2 ea
メーカー/製品名
Nalgene 383410-42
容量
10 L
10000 mL
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Specifically designed for pharmaceutical and biological manufacturing, Thermo Scientific Nalgene Polycarbonate Biotainer Bottles and Carboys reduce the risk of carry-over contamination. Durable, translucent PC construction ensures the integrity of your sample, even frozen to -100°C.
特徴および利点
- Provided sterile and ready-to-use; including certified clean products, which meet USP<788> standards and endotoxins below 0.5 EU/mL
- Certified clean products include triple layer packaging, with two heat-sealed layers to facilitate ease of entry into cleanrooms
- Sterility Assurance Level of 10-6 inside the bottle/closure
- Robust, PC can be used from -100°C to 100°C, allowing for both heated mixing and frozen storage or shipment. Must be tested in actual use conditions as results may vary depending upon application.
- Inspection procedures throughout manufacturing include leak testing, dimensional testing, visual inspection for particulates and release testing to ensure the product meets all specifications.
- All container/closure systems are certified leakproof** to protect valuable contents
法的情報
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
最新バージョンのいずれかを選択してください:
アクティブなフィルタ