表示を変更する
| サイズ/SKU | 在庫状況 | 単価 |
|---|---|---|
72 trays | カートの在庫状況を確認する | ¥186,000 |
この商品について
NACRES:
NB.22
UNSPSC Code:
41123302
Manufacturer/tradename:
Heathrow Scientific HS9902
Feature:
Holds 20 x slides 76x26 mm
Material:
black (trim)
Packaging:
case of 72 trays
¥186,000
カートの在庫状況を確認する
material
black (trim)
feature
Holds 20 x slides 76x26 mm
packaging
case of 72 trays
manufacturer/tradename
Heathrow Scientific HS9902
L × W × H
208 × 341 × 9 mm
Features and Benefits
- Easily organize slides with the index on the front cover that corresponds with the number grid inside
- Recessed slide compartments and inside covers protect specimens. Slides cannot be touched when the cover is closed, reducing contamination
- Front protection flaps are attached with durable tape for repeated use. Fold flaps flat under the slide holder to save space when in use
- Thumb cut in slide compartments enables easy removal of slides with slight pressure at one end
- Compartment holder is made from a double layer of densely pressed cardboard. Cover is a single layer cardboard
- Disposable
1 of 1
当該品目 | |||
|---|---|---|---|
| description Holds 20 x slides 76x26 mm, black (trim) | description Holds 20 x slides 76x26 mm, yellow (trim) | description Holds 20 x slides 76x26 mm, red (trim) | description Holds 30 x slides, black, trim |
| manufacturer/tradename Heathrow Scientific HS9902 | manufacturer/tradename Heathrow Scientific HS120197 | manufacturer/tradename Heathrow Scientific HS120196 | manufacturer/tradename Heathrow Scientific HS9930 |
| feature Holds 20 x slides 76x26 mm | feature Holds 20 x slides 76x26 mm | feature Holds 20 x slides 76x26 mm | feature Holds 30 x slides |
| material black (trim) | material cardboard , yellow (trim) | material cardboard , red (trim) | material cardboard |
| packaging case of 72 trays | packaging case of 72 trays | packaging case of 72 trays | packaging - |
| L × W × H 208 × 341 × 9 mm | L × W × H 208 × 341 × 9 mm | L × W × H 208 × 341 × 9 mm | L × W × H - |
Still not finding the right product?
Explore all of our products under Cardboard Slide Tray