Merck
MDE7001FS1
Millipore
Millistak+® Podデプスフィルター
DE70, filtration area 0.11m², Process Scale
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
23151806
現在、価格および在庫状況を閲覧できません。
製品名
Millistak+® Podデプスフィルター, DE70 (media series), cellulose membrane (and inorganic filter aid), effective filtration area 1.4 m2
物質
TPE (Thermo Plastic Elastomer)
TPE gasket (Thermo Plastic Elastomer)
cellulose membrane (and inorganic filter aid)
polypropylene housing (glass-filled)
品質水準
滅菌適合性
autoclavable compatible
有効ろ過面積
1.4 m2
フィッティング
seal (flat seal)
カートリッジコード
DE70 (media series)
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
The Millistak+® depth filter media is offered in a scalable, disposable format providing greater flexibility because of its unique modular design. The Millistak+® MDE7001FS1 comes in a POD format.
This DE70 media series delivers optimal performance through a gradient density matri, ideal for primary or secondary clarification applications. Millistak+® Pod filters are available in three distinct series of media grades in order to meet your specific needs - DE, CE and HC.
The Millistak+® Pod system is ideal for a wide variety of primary and secondary clarification applications, including cell cultures, yeast and E. coli lysates post centrifuge, E. coli refolds, media, vaccines, plasma proteins and sera.
Easy to Use
With the compact, modular design of our Millistak+® new Pod system, you can increase productivity and shorten cycle times.
Installation and setup of the Pod system is simple and straightforward. The unique design of the disposable adapters and disposable manifolds makes it easy to connect the Pod filters to rest of the unit operations in the process. The self-contained and disposable nature of the system protects operators from exposure to biohazards and eliminates maintenance as well as cleaning validation requirements.
特徴および利点
  • Low hold-up volume for greater product yield
  • Broad range of media types offered in single and multilayer products
  • Flexible, modular format offers scalability from 5 to 12,000 liters or more
  • Patented disposable design eliminates need for housing, CIP or cleaning validation
  • Self-contained Pod filters protect operators from exposure to biohazards
  • Robust construction is easy to use and set up
  • Smaller footprint facilitates use in tight spaces
法的情報
MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany
Emprove® プログラムとは?
Emprove®プログラムは、フィルター、シングルユースコンポーネント、医薬品原材料、出発物質について、包括的な文書を提供するシステムです。Emprove®プログラムのサブスクリプションには4種類の文書が含まれます。
Material Qualification Dossier
無料 - ログインしてダウンロード
利点について詳細を学びます。Emprove®プログラムのサブスクリプションでは、以下が提供できます。
これらの文書はなぜ閲覧不可なのですか?
Emprove®ドシエには、医薬品製造業者の登録過程やリスクアセスメント活動の支援を意図した情報及びconfidential情報が含まれています。閲覧したい場合は、お問い合わせにお問い合わせください。