おすすめの製品
由来生物
rabbit
抗体製品の状態
serum
抗体製品タイプ
primary antibodies
クローン
polyclonal
交差性
rat
メーカー/製品名
Chemicon®
テクニック
immunohistochemistry: suitable
輸送温度
dry ice
ターゲットの翻訳後修飾
unmodified
特異性
p-Tyramine
The cross-reactivities were determined using either ELISA or RIA techniques, at concentration/unconjugated or conjugated amino acid concentration at half displacement.
Compound Cross-reactivity
p-Tyramine-G-BSA 1
Octopamine-G- BSA 1/800
Dopamine-G- BSA >>40,000
Tyrosine-G- BSA 1/>50,000
The antisera was also tested for specificity using the free-floating PAP technique on rat brain.
Abbreviations:
(G) Glutaraldehyde
(BSA) Bovine Serum Albumin
The cross-reactivities were determined using either ELISA or RIA techniques, at concentration/unconjugated or conjugated amino acid concentration at half displacement.
Compound Cross-reactivity
p-Tyramine-G-BSA 1
Octopamine-G- BSA 1/800
Dopamine-G- BSA >>40,000
Tyrosine-G- BSA 1/>50,000
The antisera was also tested for specificity using the free-floating PAP technique on rat brain.
Abbreviations:
(G) Glutaraldehyde
(BSA) Bovine Serum Albumin
免疫原
p-Tyramine-gluteraldehyde-BSA
アプリケーション
Research Category
ニューロサイエンス
ニューロサイエンス
Research Sub Category
神経伝達物質及び受容体
神経伝達物質及び受容体
Detect Tyramine using this Anti-Tyramine Antibody validated for use in IH.
Immunohistochemistry: 1:1,000-1:2,500 by PAP (see suggested protocol).
Optimal working dilutions must be determined by the end user.
Optimal working dilutions must be determined by the end user.
物理的形状
Rabbit antiserum. Liquid with 0.05% sodium azide.
保管および安定性
Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
法的情報
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
免責事項
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
適切な製品が見つかりませんか。
製品選択ツール.をお試しください
保管分類コード
10 - Combustible liquids
WGK
WGK 2
引火点(°F)
Not applicable
引火点(℃)
Not applicable
適用法令
試験研究用途を考慮した関連法令を主に挙げております。化学物質以外については、一部の情報のみ提供しています。 製品を安全かつ合法的に使用することは、使用者の義務です。最新情報により修正される場合があります。WEBの反映には時間を要することがあるため、適宜SDSをご参照ください。
Jan Code
AB124:
試験成績書(COA)
製品のロット番号・バッチ番号を入力して、試験成績書(COA) を検索できます。ロット番号・バッチ番号は、製品ラベルに「Lot」または「Batch」に続いて記載されています。
Begoña Porteiro et al.
Nature communications, 8, 15111-15111 (2017-05-10)
p53 family members control several metabolic and cellular functions. The p53 ortholog p63 modulates cellular adaptations to stress and has a major role in cell maintenance and proliferation. Here we show that p63 regulates hepatic lipid metabolism. Mice with liver-specific
Irina T Sinakevitch et al.
Frontiers in systems neuroscience, 11, 77-77 (2017-11-09)
This article describes the cellular sources for tyramine and the cellular targets of tyramine via the Tyramine Receptor 1 (AmTyr1) in the olfactory learning and memory neuropils of the honey bee brain. Clusters of approximately 160 tyramine immunoreactive neurons are
Identification of distinct tyraminergic and octopaminergic neurons innervating the central complex of the desert locust, Schistocerca gregaria.
Homberg, U; Seyfarth, J; Binkle, U; Monastirioti, M; Alkema, MJ
The Journal of Comparative Neurology null
Yuki Ishikawa et al.
PloS one, 11(5), e0154230-e0154230 (2016-05-20)
In termites, i.e. a major group of eusocial insects, the soldier caste exhibits specific morphological characteristics and extremely high aggression against predators. Although the genomic background is identical to the other non-aggressive castes, they acquire the soldier-specific behavioral character during
Chia-Lin Wu et al.
Current biology : CB, 23(23), 2346-2354 (2013-11-19)
Drosophila olfactory aversive conditioning produces two components of intermediate-term memory: anesthesia-sensitive memory (ASM) and anesthesia-resistant memory (ARM). Recently, the anterior paired lateral (APL) neuron innervating the whole mushroom body (MB) has been shown to modulate ASM via gap-junctional communication in
アクティブなフィルタ
ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.
製品に関するお問い合わせはこちら(テクニカルサービス)