Merck
Z743141
Julabo® cryo-compact circulator
CF41, capacity 5.5 L, temperature -40-200 °C, precision: ±0.02, AC/DC input 115 V AC, US 3-pin plug (6-20p)
ログイン組織・契約価格を表示する
サイズを選択してください
1 EA
¥1,907,000
¥1,907,000
Check Cart for Availability
サイズを選択してください
表示を変更する
1 EA
¥1,907,000
About This Item
UNSPSCコード:
41103706
NACRES:
NB.77
¥1,907,000
Check Cart for Availability
包装
pack of 1 ea
メーカー/製品名
Julabo 9400341
温度測定範囲
-40-200 °C, precision: ±0.02
キャパシティ
5.5 L
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
The new CF series offers compact refrigerated/heating circulators. With small overall dimensions the units can even be placed on a very small surface or within a complex installation. All models offer a heating capacity of 2 kW and safety class 3 according to DIN12876-1. The maximum ambient temperature of +40°C as well as a ventilation air cooling allow installing the units very closely side by side with other units or directly under a fume hood. Furthermore all the models feature pump connections for external temperature applications and a bath opening for temperature control of small objects directly in the circulator bath. Wetted parts are made of high quality stainless steel or plastic. The instruments provide a reliable Microprocessor electronics with high temperature stability as well as warning and safety functions.
法的情報
Julabo is a registered trademark of Julabo USA, Inc.
最新バージョンのいずれかを選択してください:
試験成績書(COA)