Merck
Z741837
Extech Portable Sucrose Brix Refractometer (0 to 32%)
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41100000
NACRES:
NB.61
特徴
(WEEE compliant)
包装
pkg of 1 ea
メーカー/製品名
Extech Instruments RF10
測定範囲
0 (sucrose (Brix))
サイズ
6.6 in. × 1.6 in. × 1.2 in., 170 x 40 x 30mm
重量
7 oz200 g
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
  • Measure the concentration of Sugar, Salt, Lubricants, Antifreeze, Cutting Fluids & more
  • Easy to operate refractometers provide accurate and repeatable measurements on easy to read scales
  • Automatic temperature compensation models available for sugar, salt, lubricants, cutting fluids, and antifreeze
  • Requires only 2 or 3 drops of solution
  • The prism and lens with a simple focus adjustment provides repeatable results
  • Complete with case, calibration screwdriver, and calibration solution
最新バージョンのいずれかを選択してください:
試験成績書(COA)