Merck
Z274089
Sigma-Aldrich
Techware PP bench tray
pkg of 1 ea
ログイン組織・契約価格を表示する
サイズを選択してください
1 EA
¥41,200
¥41,200
Check Cart for Availability
サイズを選択してください
表示を変更する
1 EA
¥41,200
About This Item
UNSPSCコード:
41122800
NACRES:
NB.35
¥41,200
Check Cart for Availability
包装
pkg of 1 ea
詳細
Heavy-duty PP tray provides a solid, stable work surface. Chemically resistant and easily cleaned. Raised 1 in. lip on edges of tray provides containment of up to 2.8 L in case of spillage. Excellent for use in medium and large-size Aldrich AtmosBags as a weighted base. Built-in threaded adapter allows a 5/8 in. diam. lattice rod to be secured to tray as a vertical support for clamps. Lattice rod shown in picture not supplied with this product, purchase Z225665 separately.
最新バージョンのいずれかを選択してください:
試験成績書(COA)