Lecture-bottle gas regulators include a PTFE sealing washer. Spare PTFE sealing washers can be ordered.Product Z146994, PTFE sealing washers, CGA Inlet size 180 can be used for Products Z148504, Aldrich® corrosive lecture-bottle gas regulator, 0-15 psig delivery pressure, CGA Inlet size 180 and Z146714, Aldrich® non-corrosive lecture-bottle gas regulator, 0-15 psig delivery pressure, CGA Inlet size 180.Product Z513105, PTFE sealing washers, CGA Inlet size 170 can be used for Products Z512958, Aldrich® non-corrosive lecture-bottle gas regulator, 0-15 psig delivery pressure, CGA Inlet size 170 and Z513113, Aldrich® non-corrosive lecture-bottle gas regulator, 0-100 psig delivery pressure, CGA Inlet size 170.
¥84,600
単価¥282,000割引70%在庫あり詳細
おすすめの製品
delivery pressure measuring range
1-15 psig
CGA注入口サイズ
180
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
- Single-stage
- CGA inlet connection, needle valve outlet with ¼ in. NPTM connection
- Max. inlet pressure: 0-4000 psig
- Temp. range: -40 to 140°F
- 316 Stainless Steel body 1 ½ in. gauge face, PTFE seals and inner friction sleeves
- NPTF bonnet-vent (1/8 in.) connection for piping regulator to fume hood in case of diaphragm failure; order bonnet-vent tubing connectors separately
- Use with "T"-purge valves (listed separately) when regulating corrosive gases to minimize gas hydrolysis and corrosion of equipment
- Use for high-purity applications
Lecture-bottle gas regulators include a PTFE sealing washer. Order spare PTFE sealing washers separately.
その他情報
テクニカルインフォメーションAL-151 Gas Regulators: Selection, Installation and Operationをご覧ください。
法的情報
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
最新バージョンのいずれかを選択してください:
この製品を見ている人はこちらもチェック
-
What is the product number for the PTFE washer for the Lecture Bottle Gas Regulators?
1 回答-
役に立ちましたか?
-
-
What is the Department of Transportation shipping information for this product?
1 回答-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.