Merck
CAD6029
1-way stopcock
male Luer lock to 6 mm hose end
ログイン組織・契約価格を表示する
サイズを選択してください
1 EA
¥20,830
¥20,830
Check Cart for Availability
サイズを選択してください
表示を変更する
1 EA
¥20,830
About This Item
UNSPSCコード:
41122000
NACRES:
NB.03
¥20,830
Check Cart for Availability
包装
pack of 1 ea
メーカー/製品名
Cadence Science 6029
類似した製品をお探しですか? 訪問 製品比較ガイド
関連するカテゴリー
詳細
Stopcocks are precision-machined, individually lapped, and carefully mated to maintain pressure characteristics. These stopcocks are easily integrated with other stopcocks, manifolds, adapters, tubing, needles, and glass syringes.
  • Made from nickel plated brass or stainless steel
  • Tested to 45 psi water pressure
  • High pressure stopcocks are rated to 290 psi
最新バージョンのいずれかを選択してください:
試験成績書(COA)
文書ライブラリにアクセスする
この製品を見ている人はこちらもチェック
Slide 1 of 7
1 of 7
3-way stopcock female Luer lock to male Luer lock, left side, female Luer lock
CAD6017
3-way stopcock
3-way stopcock female Luer lock to male Luer lock, left side, female Luer lock, stainless steel
CAD6017SS
3-way stopcock
1-way tubing connector MLL to hose end for 1/4 in. to 5/16 in. I.D. tubing (316SS)
CAD6512
1-way tubing connector
3-way stopcock female Luer lock to female Luer lock, right side, female Luer lock
CAD6002
3-way stopcock
3-way stopcock female Luer lock to male Luer lock, right side, female Luer lock, compression-nut
CAD6011
3-way stopcock