Merck
CAD4045
Standard point hypodermic needles, 304 stainless steel
gauge 22, L 1 1/2 in., nickel plated hub
ログイン組織・契約価格を表示する
サイズを選択してください
12 EA
¥47,010
¥47,010
出荷予定日2025年4月27日
サイズを選択してください
表示を変更する
12 EA
¥47,010
About This Item
UNSPSCコード:
42142523
NACRES:
NB.03
¥47,010
出荷予定日2025年4月27日
物質
304 stainless steel needle
nickel plated hub
包装
pack of 12 ea
メーカー/製品名
Cadence Science 4045
L
1 1/2 in.
ゲージ
22
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Hypodermic needles are precision made and designed to ensure strict adherence with U.S. Government specification GG-N-196. Points are machine ground to achieve uniform sharpness. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure freedom from burrs and debris. All needle cannula are fabricated of type 304 stainless steel tubing which must pass rigorous quality control prior to acceptance and use.
Standard nickle plated Luer hubs comply with Luer taper specifications ISO 594. This assures an interchangeable fit mating with Luer slips and Luer locks. Contoured hub cavity facilitates cleaning.
最新バージョンのいずれかを選択してください:
試験成績書(COA)