Documenti
58806
Deburring Tool
configured for 1/8 in. tubing
Sinonimo/i:
Dressing tool
About This Item
Codice UNSPSC:
41115700
Confezionamento
pkg of 1 ea
Compatibilità
configured for 1/8 in. tubing
Descrizione generale
Deburring tool is an accessory that can be used with 1/8 in. tubing for dressing it without distortion after being smooth-cut using the tubing cutter.
Prodotti consigliati
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificati d'analisi (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Servizio Clienti