Documenti fondamentali
502472
MicroTee
for use with MicroTight Sleeves, pkg of 1 ea
About This Item
Codice UNSPSC:
41121815
Descrizione
Connects 3 pieces of 0.025″ O.D. capillary tubing
includes 3, microferrules for 0.025″ O.D. tubing and 3, female nuts (5/16"-24 internal threads)
Confezionamento
pkg of 1 ea
Compatibilità
for use with 0.025″ O.D. tubing
for use with MicroTight Sleeves
Descrizione generale
Connects 3 capillary tubes, using MicroFingertight II fittings. Includes 3 MicroFerrules for 0.025 in. O.D. tubing, 3 female nuts.
Prodotti consigliati
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)