Documenti fondamentali
Z740380
Eco-Friendly SpecTainer, tamper evident, biodegradable
size 120 mL, non-sterile
Scegli un formato
300 EA
445,00 €
445,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
300 EA
445,00 €
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.22
445,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
polyethylene cap
polypropylene container
Sterilità
non-sterile
Confezionamento
pack of 300 ea
Produttore/marchio commerciale
Simport C566-120DOECO
Misura
120 mL
Descrizione generale
The Simport Eco-Friendly SpecTainer will biodegrade to become some of the soil′s organic components in less than 7 years in constant contact with the soil instead of up to 400 years when using conventional plastics.
The Simport Eco-Friendly SpecTainer, one of the most innovative specimen containers on the market today, is now available in a new version: The Eco-Friendly SpecTainer, a rapidly degradable Specimen Container. Plastics take hundreds of years to degrade naturally in the environment. The only condition that is necessary for the Eco-Friendly SpecTainer to biodegrade is constant contact with other degrading material. No heat, physical stress, oxygen or sunlight necessary. The Eco-Friendly SpecTainer has the ability to break down safely and quickly, by biological means, into the raw materials of nature and disappear into the environment.
Does not contain heavy metals. No negative effect to its physical properties. No heat, light or mechanical stress needed for product breakdown. No toxic residue. Fully degradable within 7 years, aerobically or anaerobically.
Note legali
SpecTainer is a trademark of Simport
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)