Key Documents
Z693723
Duran® filtering flask, with Keck® assembly set
capacity 2000 mL
Sinonimo/i:
212046351
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
borosilicate glass
flat bottom flask
Giunto
joint ST/NS neck60 mm I.D.
Sterilità
non-sterile
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
DURAN® 212046351
Disponibilità
not available in USA and Canada
Capacità
2000 mL
Descrizione generale
The Duran filtering flask includes a Keck clip assembly and is idea for vacuum filtrations. The plastic hose connections meet safety and product regulations, and the flask has the Erlenmeyer shape.
Manufactured from Duran borosilicate glass type 3.3, the flasks have maximum chemical and thermal resistance and meet ISO 6556 standard.
Note legali
DURAN is a registered trademark of DWK Life Sciences
Keck is a registered trademark of Adcuram Lab Technologies GmbH