Documenti fondamentali
Z688800
Nunc® UpCell Surface cell culture multidish
size 6 wells, case of 6 ea
Sinonimo/i:
cell culture multidish, tissue culture multidish
Scegli un formato
6 EA
360,05 €
360,05 €
Prezzo di listino379,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
6 EA
360,05 €
About This Item
Codice UNSPSC:
41121812
NACRES:
NB.11
360,05 €
Prezzo di listino379,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
clear plate
clear wells
Sterilità
sterile
sterile
Caratteristiche
lid
w/ lid and airvent
w/ membrane
Confezionamento
case of 6 ea
Produttore/marchio commerciale
Nunc 174901
Lungh. × Largh. vaschetta
128 mm × 86 mm
Lungh. × largh. esterne
128 mm × 86 mm
Misura
6 wells
Area superficiale
9.6 cm2 , culture area (cm2)/well
Volume d’esercizio
3 mL
Tipo di binding
(UpCell Surface)
Descrizione generale
The Nunc UpCell Surface dishes have a temperature responsive cell culture surface. These UpCell Surface dishes are suitable for harvesting of cells with intact surface proteins, for culture passaging, single-cell analyses and cell transplantation research. The UpCell surface enables harvesting of cell sheets and creation of 3D tissue models which are held together by normal cell junctions and extracellular matrix.
  • Harvesting of cells with intact surface proteins
  • No trypsinization - preserve cell surface proteins
  • No physical force - get high cell viability
  • Releases adherent cells by reduction of temperature of the cell culture
  • For culture passaging, single-cell analyses and cell transplantation research
  • Enables harvesting of cell sheets and creations of 3D tissue models held together by normal cell junctions and extracellular matrix
  • Minimal hands-on time
  • For research and single-use only
  • It is quick, clean and simple-just reduce the temperature
Note: Suitable for the harvesting of cells with intact surface proteins, for cell culture passage, single-cell analyses and cell transplantation research.
Note legali
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
UpCell is a trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)