Documenti fondamentali
Z615080
Vivapore 5 solvent absorption concentrator
MWCO 7.5 kDa
About This Item
Codice UNSPSC:
41104916
NACRES:
NB.24
Materiali
polyethersulfone membrane
styrene-acrylonitrile (SAN) housing
Descrizione
min concentrate vol 50 μL
Sterilità
non-sterile
Produttore/marchio commerciale
Sartorius VP0502
Disponibilità
not available in USA and Canada, ROW
Area membrana
20 cm2
Volume d’esercizio
5 mL
Dimensione pori
7.5 kDa MWCO
Descrizione generale
With no need for additional equipment, pressure or vacuum, solvent absorption is the most economic and user friendly concentration technique available to the clinician and research scientist. Just fill the unit with the solution to be concentrated, wait for the desired concentration level to be achieved and then pipette the concentration sample from the bottom of the reservoir. Ideal for general purpose laboratory concentration or purification prior to further analysis, Vivapore is particularly suited for labile solutions that can denature with alternative shear or pressure inducing methods or require processing in a cold room environment. Vivapore concentrators extend the solvent absorption technique to a totally new level of performance, application potential and ease of use. Requires stand.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)