Documenti fondamentali
TMO156545
Nunc® MicroWell 96-well Plate with high flange skirt
Nunclon® Delta surface, polystyrene, with lid, clear, case of 180 ea
Sinonimo/i:
multiwell plate, non treated multiwell plate, non treated plate
Scegli un formato
180 EACH
1.010,00 €
1.010,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
180 EACH
1.010,00 €
About This Item
Codice UNSPSC:
41122102
NACRES:
NB.15
1.010,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
clear
polystyrene
Confezionamento
case of 180 ea
Produttore/marchio commerciale
Nunc 156545
Descrizione generale
Obtain consistent and reliable results with Thermo Scientific Nunc MicroWell 96-Well Microplates. With multiple surface modifications, these plates are ideal for cell-based assays as well as biochemical assays. The flat bottom shape is suitable for microscopic and optical measurements. Available in either the high flange skirt type or the low flange pinchbar design, the plates are optimized for a range of robotics, automated readers and liquid handling systems.
Note legali
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)