Cambia visualizzazione
| A voi/SKU | Disponibilità | Prezzo |
|---|---|---|
40 trays | Per informazioni sulla disponibilità, contatta il Servizio Clienti. | 526,00 € |
Informazioni su questo articolo
NACRES:
NB.35
UNSPSC Code:
41123302
Manufacturer/tradename:
Heathrow Scientific HS9930
Feature:
Holds 30 x slides
Material:
cardboard
526,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutartimaterial
cardboard
feature
Holds 30 x slides
manufacturer/tradename
Heathrow Scientific HS9930
size
281x328x10 mm
color
black, trim
Features and Benefits
- Easily organize slides with the index on the front cover that corresponds with the number grid inside
- Recessed slide compartments and inside covers protect specimens. Slides cannot be touched when the cover is closed, reducing contamination
- Front protection flaps are attached with durable tape for repeated use. Fold flaps flat under the slide holder to save space when in use
- Thumb cut in slide compartments enables easy removal of slides with slight pressure at one end
- Compartment holder is made from a double layer of densely pressed cardboard. Cover is a single layer cardboard
- Disposable
1 of 1
Questo articolo | |||
|---|---|---|---|
| description Holds 30 x slides, black, trim | description Holds 20 x slides, blue, trim | description Holds 20 x slides, green, trim | description Holds 20 x slides 76x26 mm, black (trim) |
| feature Holds 30 x slides | feature Holds 20 x slides | feature Holds 20 x slides | feature Holds 20 x slides 76x26 mm |
| manufacturer/tradename Heathrow Scientific HS9930 | manufacturer/tradename Heathrow Scientific HS9913 | manufacturer/tradename Heathrow Scientific HS9914 | manufacturer/tradename Heathrow Scientific HS9902 |
| material cardboard | material cardboard | material cardboard | material black (trim) |
| color black, trim | color blue, trim | color green, trim | color - |
| size 281x328x10 mm | size 208x341x9 mm | size 208x341x9 mm | size - |
Still not finding the right product?
Explore all of our products under Cardboard Slide Tray
Scegli una delle versioni più recenti:
Possiedi già questo prodotto?