Documenti fondamentali
HS120255
Wide mouth safety labelled wash bottle
capacity 500 mL, label, Sodium hypochlorite, self venting, PE-LD
Scegli un formato
1 EACH
56,00 €
56,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EACH
56,00 €
About This Item
Codice UNSPSC:
41103209
NACRES:
NB.43
56,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
PE-LD
blue bottle
high-density polyethylene cap
low-density polyethylene bottle
wide-mouth bottle
Caratteristiche
label Sodium hypochlorite
vented
with cap
Confezionamento
pack of 6 ea
Produttore/marchio commerciale
Heathrow Scientific HS120255
Capacità
500 mL
Descrizione generale
Clear and informative product safety information.
The information provided on the front of each bottle is specific to the product contained and includes the following — the Chemical Abstract Service (CAS) No., National Fire Protection Association (NFPA) codes and symbols, Protective Equipment Symbol (PES), Target Organ Information and Hazard Codes. You also have the option of using other liquids within the bottles and creating the appropriate safety information on the outside of the bottle by using the ‘Self Labeling′ versions. The closure and stem are molded in one piece to make them leak-proof. The angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow.
  • Wide bottle mouth makes filling easier
  • Non-metallic vent that prevents the risk of solvent leakage
  • With the exception of the bottle designed for Sodium Hypochlorite, which is a white LDPE bottle for light protection and has a HDPE closure, the bottles are made from LDPE, with a PP closure
  • Six named chemicals, a self labeling and assorted pack of bottles to choose from
  • 500 mL bottle size
  • Supplied in packs of 6
Self Labeling Bottles must only be used with chemicals that are safe and compatible with LDPE
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)