Documenti fondamentali
GEUS77720
illustra ExoProStar 1-Step
2000 reactions
Sinonimo/i:
ExoProStar 1-Step
Scegli un formato
1 EACH
1.580,00 €
1.580,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EACH
1.580,00 €
About This Item
Codice UNSPSC:
41121800
NACRES:
NA.52
1.580,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Descrizione generale
Enzymatic PCR clean up technology with illustra Exonuclease I and Alkaline Phosphatase combined in one tube to remove unincorporated primers and dNTPs. For maximum convenience comes in a single tube.
illustra ExoProStar is our new name for this PCR clean-up technology. No change in the formulation, so you get the same great product, and same great benefits!
The ExoProStar 1-Step PCR and Sequence Reaction Clean-Up reagent provides a simple, single-step method for removing unincorporated primers and nucleotides prior to downstream analysis. illustra Exonuclease I and Alkaline Phosphatase enzymes are combined in a single, stable mix requiring just one pipetting step to set up the clean-up reaction.
illustra ExoProStar 1-Step is based on GE′s founding patents for the clean-up of PCR reactions using Exonuclease 1 and Alkaline Phosphatase. This 1-step version is not available in USA, Canada and Japan. Instead, we offer a version with the enzymes in two separate tubes, the ExoProStar.
Caratteristiche e vantaggi
  • One tube with one mix of enzymes delivering PCR and sequence reaction clean-up
  • Just one pipetting step to set up your reaction
  • Fast 30-minute, hands-free protocol
  • 100% recovery of PCR product
  • Scalable for different reaction sizes
  • Automation friendly
Note legali
illustra is a trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)