Documenti fondamentali
GE28-9889-47
Sigma-Aldrich
XK 16/100 Column
Scegli un formato
1 EA
952,00 €
952,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EA
952,00 €
About This Item
Codice UNSPSC:
41115700
NACRES:
SB.52
952,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
polypropylene column
Confezionamento
1 ea of
Descrizione generale
XK empty columns are designed for liquid chromatography at low to medium pressure. The user-friendly design ensures trouble-free operation with fittings for direct connection to ÄKTA design instruments or other high-performance liquid chromatography systems. The product family is based on a robust design for high reproducibility and accurate results. XK columns are compatible with aqueous solutions and most organic solvents used in liquid chromatography of macromolecules.
Caratteristiche e vantaggi
Adapter QuickLock mechanism that facilitates column handling and cleaning
Cooling jacket for sensitive purifications
Fittings for direct connection to chromatographiy systems
Plunger design to ensure a homogeneous buffer distribution and reproducible packing
User friendly and robust empty columns, designed for liquid chromatography at low to medium pressure.
Prodotti consigliati
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Protocolli
Column Packing and Preparation for Size Exclusion Chromatography