Documenti fondamentali
GE28-9322-23
Vivaspin® 500, 5 kDa MWCO Polyethersulfone
Cytiva 28-9322-23, pack of 25 pieces
Scegli un formato
25 EA
139,00 €
139,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
25 EA
139,00 €
About This Item
Codice UNSPSC:
41104916
NACRES:
NB.24
139,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
polycarbonate housing
polyethersulfone membrane
Sterilità
non-sterile
Confezionamento
pack of 25 pieces
Produttore/marchio commerciale
Cytiva 28-9322-23
Disponibilità
not available in Japan
Dimens. tubo
~2.2 mL
Volume d’esercizio
500 μL
Dimensione pori
5 kDa MWCO
Temperatura di conservazione
room temp
Descrizione generale
Designed for fast, nondenaturing concentration of biological samples by membrane ultrafiltration. Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%.
Vivaspin® sample concentrators are designed for fast, nondenaturing concentration of biological samples by membrane ultrafiltration. Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. The entire process is performed in a single tube with an upper compartment containing sample and lower compartment separated by a semipermeable membrane with a molecular weight cut-off (MWCO) selected by the user. Centrifugation is applied to force solvent through the membrane, leaving a more concentrated sample in the upper chamber.
Vivaspin® sample concentrators cater for sample volumes from 100 μl to 20 ml, with a range of molecular weight cutoff values from Mr 3 000 to 100 000. For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest
Caratteristiche e vantaggi
  • Compatible pH range is from pH 1 to 9.
  • Patented dead-stop technology, which ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery
  • One-step sample concentration in a single tube for minimal sample handling and reduced sample loss.
  • Vertical polyethersulfone membrane minimizes membrane blockage and tolerates high flow rates.
Stoccaggio e stabilità
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Risultati analitici
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Note legali
Vivaspin is a registered trademark of Sartorius Stedim Biotech GmbH
Codice della classe di stoccaggio
13 - Non Combustible Solids
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Articoli
This page describes troubleshooting strategies in purification of GST-tagged proteins using Cytiva products.
Protocolli
Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.
This page shows how to perform sample desalting, buffer exchange and concentration for affinity chromatography of tagged proteins.