Documenti fondamentali
GBL705016
Grace Bio-Labs ONCYTE® SuperNOVA nitrocellulose film slides
16-6 mm x 6 mm NC pads per slide, plain glass
Sinonimo/i:
nitrocellulose slides, protein microarrays
Scegli un formato
20 EA
892,00 €
892,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
20 EA
892,00 €
About This Item
Codice UNSPSC:
41122600
NACRES:
NB.15
892,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
nitrocellulose (on glass)
Confezionamento
pkg of 20 ea
Produttore/marchio commerciale
Grace Bio-Labs 705016
Dimens. esterne
25 mm × 75 mm
Tipo di binding
high binding surface
Descrizione generale
Fluorescence detection is a preferred method for many quantitative protein microarray assaysin research and diagnostic applications due to its excellent sensitivity and potential for multiplexing. Use of multiplexed fluorescence-based assays on porous nitrocellulose though has been hampered by high backgrounds at shorter excitation wave lengths (such as the 532nm fluorescence channel). These ONCYTE™ SuperNOVA™ microporous nitrocellulose film slides increase protein microarray assay sensitivity and dynamic range using fluorescent end points at multiple wave lengths. Use of SuperNOVA™ slides can lower the limit of detection with fluorescent end points by 3 – 4 orders of magnitude over competing nitrocellulose film slides.
Note legali
NOVA is a trademark of Grace Bio-Labs, Inc.
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
SuperNOVA is a trademark of Grace Bio-Labs, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)