Documenti fondamentali
GBL665301
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side
(Adhesive on one side)
Sinonimo/i:
Nonadhesive reagent isolators, autoclavable slide chambers, removable hydrophobic barrier
Scegli un formato
25 EA
220,00 €
220,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
25 EA
220,00 €
About This Item
Codice UNSPSC:
41161502
NACRES:
NB.13
220,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
red silicon
Caratteristiche
(Adhesive on one side)
Confezionamento
pack of 25 ea
Produttore/marchio commerciale
Grace Bio-Labs 665301
Lungh. × largh. esterne
51 mm × 25 mm
Superficie del pozzetto
0.66 cm2
Pozzetti
8
Descrizione generale
Silicone Isolators allow researchers to isolate specimens using removable hydrophobic barriers. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides. Isolators are available in red silicone thickness 0.5 to 2.5mm pre-cut or as sheet material with or without secure seal clean adhesive on one or both surfaces. Nonadhesive isolators are autoclavable and reusable.
Silicone Isolators remain sealed to smooth surfaces during washing steps. Where additional sealing is required, SecureSeal adhesive on one or both surfaces is recommended. Each seal design is also available in CultureWell "water-clear" silicone, which is highly recommended for sensitive cell culture applications. Closed chambers may be formed using flexible RNase free, HybriSlip covers. Recommended use temperature is -62°C to 218°C.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)