Documenti fondamentali
GBL470638
Grace Bio-Labs ProPlate® microarray system, slide module only
well size 8, L × W × depth 6 mm × 20 mm × 7.6 mm, without clips
Sinonimo/i:
incubation chambers, microarray hybridization chambers, slide chambers
Scegli un formato
10 EA
234,00 €
234,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
10 EA
234,00 €
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
234,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
black silicone (on Delrin)
Caratteristiche
without clips
Confezionamento
pack of 10 ea
Produttore/marchio commerciale
Grace Bio-Labs 470638
Lungh. × largh. × prof.
6 mm × 20 mm × 7.6 mm
Dimensione pozzetto
8
Descrizione generale
Grace Bio-Labs ProPlate® multi-array chambers (microarray systems) are designed to be used with ONCYTE® nitrocellulose-coated slides. These multi-array chambers integrate microscope slide based microarray technology with high-throughput (HTS applications) microtiter plate processing. Up to 64 individual arrays can be assayed per slide.
Three or four slides fit into a tray (depending on clip choice). The resulting multi-array chamber tray set produces a modular plate with a standard microtiter plate footprint and well spacing allowing high-throughput robotic processing of up to 256 arrays.
Features:
  • Up to 64 leak-proof wells on any slide surface without the use of adhesive
  • Microtiter plate well spacing facilitates use with multichannel pipettes
  • 7 x 7 mm square wells are ideal for high content arrays 5-350 ul well volumes accommodate small reagent and large wash volumes
Note legali
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
ProPlate is a registered trademark of Grace Bio-Labs, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)