Documenti fondamentali
EP3124000067
Eppendorf Research® plus Pipette, fixed volume
single-channel, volume 50 μL, yellow
Scegli un formato
1 EA
199,00 €
199,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EA
199,00 €
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
199,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
yellow
Caratteristiche
single-channel
mechanical
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Eppendorf® 3121000066 (Global)
Volume
50 μL
Descrizione generale
The Eppendorf Research plus mechanical pipette is the result of over 60 years of experience in liquid handling and is one of the most frequently used pipettes in the world. Thanks to the groundbreaking Eppendorf PhysioCare Concept, the Research plus is very ergonomic and protects your health in the laboratory. It is characterized by low weight and low operating forces, is extremely reliable, fully autoclavable and easy to use. It feels good to know that you are doing the right thing – knowing that you are working with one of the most well-known and advanced pipettes in the world. A spring-loaded tip cone, an option for temporary adjustment, an improved volume display – and all this in a lightweight, fully autoclavable pipette: that is the Eppendorf Research plus manual pipette. In 2021, the Research plus single-channel variable volume pipettes were certified with the ACT Environmental Impact Factor Label by the non-profit organization My Green Lab . This makes them a good choice for laboratories looking for more sustainable products.
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Research is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)