Documenti
EP022638653
Wide Neck Bottle for Eppendorf® A-4-81 Rotor
round, capacity 400 mL, pack of 2
About This Item
Codice UNSPSC:
41121703
Materiali
polypropylene bottle
round
Caratteristiche
with cap
Confezionamento
pack of 2
Produttore/marchio commerciale
Eppendorf® 022638653 (US)
Parametri
3250 × g max. RCF
Capacità
400 mL
Categorie correlate
Note legali
Eppendorf is a registered trademark of Eppendorf AG