Documenti fondamentali
EP0030078861
Eppendorf® ep Dualfilter T.I.P.S.
PCR clean, sterile, working volume 5-100 μL, L 53 mm, yellow, pack of 3840 ea (10 racks of 384 tips)
Sinonimo/i:
Eppendorf® ep Dualfilter T.I.P.S.®, pipette tip
Scegli un formato
3840 EA
918,00 €
918,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
3840 EA
918,00 €
About This Item
Codice UNSPSC:
41121600
NACRES:
NB.15
918,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
yellow
Sterilità
sterile
Caratteristiche
PCR clean
Confezionamento
pack of 3840 ea (10 racks of 384 tips)
Produttore/marchio commerciale
Eppendorf® 0030078861
Lunghezza
53 mm
Volume
100 μL
Volume d’esercizio
5-100 μL
Categorie correlate
Descrizione generale
Unique two-phase filter protection with ep Dualfilter T.I.P.S. The Eppendorf ep Dualfilter T.I.P.S. are the first filtertips with a two phase filter for contamination protection. The tips are manufactured to the highest possible quality from pure, non recycled materials under cleanroom conditions. They are characterized by defined flow dynamics, low wettability and high thermal stability. They are certified pyrogen free and PCR clean (free from human DNA, DNase, RNase and PCR-inhibiting substances). These ep Dualfilter T.I.P.S. contain well-defined different pore sizes within the two filter layers. The white layer which faces the sample retains drops, splashes and aerosols. The blue layer above it forms a highly-effective double barrier, that reliably binds even biomolecules. Without any kind of filter the pipette is exposed to contamination by sample and aerosols. Conventional single layer filters do not fully block particles and molecules. Only ep Dualfilter T.I.P.S. provide reliable protection against even the finest impurities.
Caratteristiche e vantaggi
  • Two filter layers provide dual protection for pipette and sample
  • Two filter layers provide dual protection from aerosols and biomolocules
  • Free from PCR inhibitor additives
  • Patent pending technology
  • All tips are provided sterile, Eppendorf PCR clean and pyrogen-freeBatch certified
  • Defined flow dynamics
  • Low wettability
  • High thermal stability
  • Leak proof seal
  • Minimum attachment and ejection forces
Note legali
Eppendorf is a registered trademark of Eppendorf AG
ep Dualfilter T.I.P.S. is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)