Documenti fondamentali
CLS431841
Corning® centrifuge bottle
with standard screw cap, capacity 250 mL, polypropylene
Sinonimo/i:
centrifugation bottles, digestion bottles, shaking bottles
About This Item
Codice UNSPSC:
41121703
NACRES:
NB.11
Materiali
flat bottom tube
polypropylene
round bottle
translucent bottle
wide-mouth bottle
Sterilità
non-sterile
Caratteristiche
autoclavable
cap
Confezionamento
pkg of 36 ea
Produttore/marchio commerciale
Corning 431841
Parametri
13500 × g max. RCF
Capacità
250 mL
Caratteristiche e vantaggi
  • Corning® centrifuge bottles have a specially designed cap for improved grip
  • Includes clear graduations and textured marking spot.
  • Bottles are designed with extra-thick walls for high durability
  • Wide mouth for easier liquid filling
  • Compatible with most centrifuges and do not require special buckets
  • Unique design aid researchers to reduce the number of centrifugation cycles and enhance productivity
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)