Documenti fondamentali
ZWDK6L100
Milli-Q
Kit per la distribuzione di acqua alle lavavetrerie
AC/DC input 115 V, left, Cost-effective solution to feed high-quality pressurized pure water to laboratory appliances
Scegli un formato
1 KIT
1.310,00 €
1.310,00 €
Spedizione prevista il17 febbraio 2025Dettagli
Richiedi maggiori informazioni
Scegli un formato
Cambia visualizzazione
1 KIT
1.310,00 €
About This Item
Codice UNSPSC:
41104200
NACRES:
JA.13
1.310,00 €
Spedizione prevista il17 febbraio 2025Dettagli
Richiedi maggiori informazioni
impiego
(Compatible with storage tanks 50 L 60 L and 100 L)
Confezionamento
pkg of 1 unit
Produttore/marchio commerciale
Milli-Q®
Parametri
≤15 L/min distribution flow rate (for 115 V pump)
16.2 L/min flow rate at 15 psi (1 bar) (for 115 V pump)
Alt. × largh. × prof.
15.98 cm (6.3 in.) × 27.5 cm (10.8 in.) × 31 cm (12.2 in.)
Compatibilità
for use with Elix® Advantage
for use with Elix® Essential UV
for use with Elix® Essential
for use with Elix® Reference
for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015
for use with Milli-Q® Direct
for use with RiOs Essential
Descrizione generale
  • This Washer Distribution Kit is a cost-effective, pump-based solution designed to supply your laboratory appliances with a constant source of high-quality pressurized pure water.
  • The kit′s compact, smooth design allows easy installation wherever it suits you (wall-mounted, bench or under bench installation).
  • This left-side version is for installation on the left side of your reservoir as it has the water inlet on its right and a pressurized water outlet on the left.
Applicazioni
The Washer Distribution Kit will provide a silent, automatic supply of pure water when required.
Caratteristiche e vantaggi
  • Cost-effective solution ensuring pressurized pure water feed to common laboratory appliances with flow rates between 16.2 L/min @ 15 psi (1 bar)
  • Small footprint.
  • Silent operation.
  • Automatic Start/Stop of water flow to appliance ensured by an integrated pressure switch.
Note legali
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany