Documenti
Z759325
Sigma-Aldrich
Joint clip, standard taper
joint: 29, red polyethylene
Sinonimo/i:
NS joint clamps, NS joint clips, National standard joint clamps, National standard joint clips
About This Item
Codice UNSPSC:
41122413
NACRES:
NB.35
Materiali
red polyethylene
Giunto
joint: 29
Confezionamento
pkg of 50 ea
Descrizione generale
Color coded, rigid polyethylene joint clips fit standard taper joints. Will not scratch glassware or corrode; withstands temperatures up to 120 °C.
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 2
1 of 2
Plastic Clip for use with 24/40 
Supelco
64764
Plastic Clip
Keck® Joint Clip joint size, 35, ball-joint
Z222488
Keck® Joint Clip