The reservoir identified as Z680354 is the single-well reservoir, as depicted in the picture, which is the middle reservoir containing blue liquid
Z680354
Bio-Pure™ pipetting reservoir
capacity 25 mL
Sinonimo/i:
multichannel pipette reservoir, reagent reservoir
Scegli un formato
108,00 €
Scegli un formato
About This Item
108,00 €
Prodotti consigliati
Materiali
PET (Bio-degradable)
V-bottom bottom
white
Sterilità
sterile
Confezionamento
pack of 100 ea
Produttore/marchio commerciale
Diversified Biotech REBP-3000
Capacità
25 mL
Descrizione generale
Caratteristiche e vantaggi
- Constructed from biodegradable PET material for environmentally conscious disposal.
- Specialized "trough within a trough" design for optimal reagent recovery.
- Pour-off spouts at all four corners and a wide base for improved stability and spill prevention.
- Sterilized for safe and contamination-free usage.
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
-
Which of the four reservoir types displayed on the product page corresponds to Z680354?
1 risposta-