This product is standardized to a NIST standard. A Traceability Certificate is supplied with the item. A Calibration Report is not included.
Z676403
Double-Safe™ PTFE coated thermometer
L 200 mm, temperature -10-110 °C, immersion level, total
Scegli un formato
56,50 €
Scegli un formato
About This Item
56,50 €
Prodotti consigliati
Caratteristiche
immersion level total
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
HB Instruments B60400-1200
Caratteristiche più verdi
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
Campo di misura della temperatura
-10-110 °C
Lunghezza
200 mm
Categoria alternativa più verde
Categorie correlate
Descrizione generale
- PTFE coating contains liquid and glass in case of breakage
- Non-toxic, non-hazardous, biodegradable black Enviro-Safe liquid
- Available with statement of accuracy traceable to NIST and DKD
- Lead-free glass and recyclable packaging
Note legali
Prodotti correlati
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
-
Does it come with calibration certificate.
1 risposta-