Documenti fondamentali
Z648191
STATMARK permanent marker for cassette & slide marking
STATMARK permanent marker for cassette & slide marking
pkg of 12 ea
Sinonimo/i:
solvent resistant pen
Scegli un formato
12 EA
106,00 €
106,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
12 EA
106,00 €
About This Item
Codice UNSPSC:
41181600
NACRES:
MA.01
106,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Confezionamento
pkg of 12 ea
Produttore/marchio commerciale
Ted Pella 22313
Descrizione generale
The STATMARK Permanent ink pen in an ultra fine tip is ideal for labeling microscope slides and cassettes. Made of rigid nylon that allow for precise hand marking on glass slides as well as on most tissue embedding cassettes.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 1
1 of 1
Domande
1–4 di 4 domande  
  1. Is this marker solvent resistant to chloroform or low concentration acid, like 10% acetic acid?
    1 risposta
    1. When allowed to dry, the marker is resistant chemicals and solvents. Some fading may occur with exposure to ethanol or ethanol/xylene mixtures. See the statement below from the manufacturer.
      Utile?
  2. This marker writing, is stable up to 800 degree centigrade.
    1 risposta
    1. This marker has not been tested in high temperature. There is no heat resistant marker product currently available.
      Utile?
  3. Is this Statmark marker suitable for labeling tubes with a storage temperature of -80°C?
    1 risposta
    1. This marker is not rated for cryogenic temperatures. See the link below to the Nalgene Cyroware marker set, which are suitable for ultra-low temperatures: https://www.sigmaaldrich.com/product/sigma/m6163.
      Utile?
  4. is this marker solvent resistant (including formalin, xylene, and alcohols)?
    1 risposta
    1. The STATMARK permanent marker contains quick drying ink is highly resistant to aqueous based stains, alcohols, xylene and xylene substitutes.
      Utile?
Recensioni
Filtri attivi
  1. 1 recensione solo con voto