Documenti fondamentali
Z544493
Molymod® molecular model set
teacher/researcher set, organic stereochemistry
Scegli un formato
1 SET
92,60 €
92,60 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 SET
92,60 €
About This Item
Codice UNSPSC:
60104506
NACRES:
NB.17
92,60 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Caratteristiche
no. of atoms 168
no. of bonds 192
Produttore/marchio commerciale
Molymod MMS-050
Descrizione generale
166 atom-parts
60 molydomes hydrogen
44 Carbon -black ( 2 different types)
1 group alkene
1 group alkyne
16 Oxygen - red ( 3 different types)
4 Hydrogen - white
12 Nitrogen - blue ( 2 different types)
12 Sulphur - yellow ( 2 different types)
5 Phosphorus - purple ( 2 different types)
1 Metal - grey
8 chlorine
2 bromine
2 iodine
6 p-lobes pink
6 p-lobes purple
40 links, gray, medium ML-12
12 links, gray, flexible long ML-13
60 short colorles
60 short white
20 V-links
2 link remover tool
Top quality Molymod molecular model sets are for use by students, teachers, and researchers. May be upgraded or customized with additional model parts, for more info, go to sigma-aldrich.com and enter Molymod in our search engine.
Scale is 2.8 cm =100 pm (open style) and 1.45 cm = 100 pm (compact, semi-space-filling). Sets include standard color-coded plastic atom centers (with bond holes set at the appropriate angles), bonding links, and a plastic storage box.
Set pictured is representative of the Student set.
See our Selection Guide for detailed descriptions of set contents.
Note legali
Molymod is a registered trademark of Spiring Enterprises Ltd.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 2
1 of 2