Z544469
Molymod® molecular model set
teacher/researcher set, organic/inorganic
Scegli un formato
60,00 €
Scegli un formato
About This Item
60,00 €
Prodotti consigliati
Caratteristiche
no. of atoms 106
no. of bonds 86
Produttore/marchio commerciale
Molymod MMS-004
Descrizione generale
(22) red oxygen atom parts: (16) angular divalent, (6) tetrahedral
(14) white hydrogen atom parts: (12) monovalent, (2) linear divalent
(13) yellow sulfur atom parts: (8) angular divalent, (4) tetrahedral, (1) octahedral
(10) blue nitrogen atom parts: (6) tetrahedral, (4) pyramidal
(7) purple phosphorus atom parts: (4) tetrahedral, (1) trigonal bipyramidal, (2) pyramidal
(8) green monovalent halogen atom parts
(14) grey metal atom parts: (4) monovalent, (2) angular divalent, (2) trigonal planar, (4) tetrahedral, (1) octahedral
(12) Lone pair 2D paddles: (6) pink, (6) purple
(6) beige lone pair electrons
(38) medium-length 31mm grey links
(36) long flexible 46mm grey links
(12) medium-length 31mm purple links
Storage box
Instructions
Scale is 2.8 cm =100 pm (open style) and 1.45 cm = 100 pm (compact, semi-space-filling). Sets include standard color-coded plastic atom centers (with bond holes set at the appropriate angles), bonding links, and a plastic storage box.
Set pictured is representative of the Student set.
See our Selection Guide for detailed descriptions of set contents.
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)