Z515973
Aldrich® solvent storage flask
capacity 1,000 mL, joint: ST/NS 29/32
Sinonimo/i:
bottom flask, flask round, round flask
About This Item
Prodotti consigliati
Materiali
borosilicate glass
round bottom
single-neck
Giunto
joint: ST/NS29/32
Sterilità
non-sterile
Caratteristiche
closure type Standard Taper Joint
Confezionamento
pack of 1
Produttore/marchio commerciale
Sigma-Aldrich
Capacità
1,000 mL
Descrizione generale
With ground-glass joint and glass stopcock. Use septum Z565695 or Z565679 (see Septa Section).
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC