Documenti
Z302538
Quickfit® Multiple Adapters
top female Joints: ST/NS 19/26 (2), bottom male joint: ST/NS 24/29
Sinonimo/i:
MA1/3
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Giunto
female Joints: ST/NS top19/26 (2)
male joint: ST/NS bottom24/29
Produttore/marchio commerciale
Quickfit MA 1/3
Disponibilità
not available in USA and Canada
not available in USA and Canada
Descrizione generale
Two neck parallel.
Note legali
Quickfit is a registered trademark of SciLabware Limited