Documenti
Z283282
Sigma-Aldrich
Aldrich® heavy-duty spatula
L 18 in.
About This Item
Codice UNSPSC:
41122403
NACRES:
NB.35
Materiali
stainless steel
Lunghezza
18 in.
Descrizione generale
Double flat ends; one rounded, one square. Each end is 3 L × 3/4 in. W.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 7
1 of 7
Micro spatulas one scraper/spoon end, L 6 1/2 in. (165 mm)
Z193216
Micro spatulas
Lab spoons nickel-stainless steel, L 9 in.
Sigma-Aldrich
Z282774
Lab spoons
Lab Spatula Flat end and spoon end, nickel-stainless steel (plated), L 225 mm
HS15909
Lab Spatula
Micro spatulas Curved at one end with knob at the other end, L 5 1/12 in. (140 mm)
Z193208
Micro spatulas