Documenti
Z243213
Micro spatula
L 6 5/8 in., stainless steel, rod diam. 0.09 in.
Sinonimo/i:
laboratory spoon, micro spoon, small sample spoon
About This Item
Codice UNSPSC:
41122403
NACRES:
NB.35
Materiali
stainless steel
Confezionamento
pack of 2 ea
Lunghezza
6 5/8 in.
Diam. barra
0.09 in.
Descrizione generale
Bent tip and spoon. Bent sharpened tip (3 1/2 mm wide) at one end. With V-Shaped sharpened spoon at the other end. Overall length: 6 5/8" (170 mm).
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 3
1 of 3
PTFE-coated spatulas Tapered to 1/8in., L 7 1/4 in.
Z115282
PTFE-coated spatulas
Micro spatulas flat rounded/tapered PTFE coated ends, L 7 5/8 in. (190 mm)
Z513334
Micro spatulas