Documenti
Z202827
Sigma-Aldrich
Glass spatulas
Spoon and paddle ends, diam. × L 7 mm × 200 mm
About This Item
Codice UNSPSC:
41122403
NACRES:
NB.35
diam. × lung.
7 mm × 200 mm
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 6
1 of 6
Lab Spatula Flat end and spoon end, nickel-stainless steel (plated), L 225 mm
HS15909
Lab Spatula
Micro spatula L 6 5/8 in., stainless steel, rod diam. 0.09 in.
Z243213
Micro spatula
PTFE-coated spatulas Square-cut, L 7 1/4 in.
Z115274
PTFE-coated spatulas