Documenti
Z131253
Nalgene® volumetric flask
volume 100 mL, accuracy: 0.16 mL, translucent polypropylene copolymer (PPCO)
Sinonimo/i:
nalgene, plastic flask, plastic flasks
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
long
translucent polypropylene copolymer (PPCO)
Giunto
joint threaded neck20mm
Sterilità
non-sterile
Caratteristiche
accuracy class B (ASTM E288 and ISO Standard 384)
closure type Screw cap
Confezionamento
pack of 1
Produttore/marchio commerciale
Nalgene® 4000-0100
Campo di misura del volume
100 mL, accuracy: 0.16 mL
Capacità
100 mL
Descrizione generale
Gravimetrically calibrated TC/TD at 20°C individually, then marked with a permanent color graduation line. Meet Class B accuracy requirements of ASTM E288 and ISO Standard 384. No meniscus to confuse readings. Oval marking spot. Linerless PP screw closure. Leakproof and autoclavable.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 2
1 of 2