Z118133
Glass sample holder
Tube, size 2 1/2 in. × 7 in.
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.43
Prodotti consigliati
Materiali
borosilicate glass
Parametri
230-490 °C temp. limit
Misura
2 1/2 in. × 7 in.
Descrizione generale
These sample holders come in both boat and tube designs, made of borosilicate glass. The glass vessel is ideal for transporting and drying chemical samples in the Aldrich Abderhalden apparatus, in desiccators, or in any drying oven. It has a molded end piece to grip and easier handling of the boat.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)