Documenti
Z115630
Sigma-Aldrich
Aldrich® vacuum-filtration adapter
male joint: ST/NS 24/40, flange I.D. 38 mm
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Giunto
male joint: ST/NS24/40
Diam. int. flangia
38 mm
Categorie correlate
Descrizione generale
Enables filtration (via a Buchner funnel) directly into a single-neck, round-bottom flask. Includes rubber sleeve.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Ricambio