WHAWB100039
Unicore Punch Kit
I.D. 3.0 mm, pkg of 4 ea
Sinonimo/i:
UniCore Punch Kit 3.00 mm (4), Whatman FTA, Z747424, cutting mat, harris punch, harris uni-core, uni-core, whatman punch, Harris Uni-Core
Scegli un formato
74,70 €
Scegli un formato
About This Item
74,70 €
Prodotti consigliati
Confezionamento
pkg of 4 ea
Produttore/marchio commerciale
Qiagen WB100039
Diam. int.
3.0 mm
Descrizione generale
A cutting mat (also available separately: WHAWB100022) included in UniCore Punch Kits ensures clean sample cuts from FTA™/FTA™ Elute Cards and extends the life time of the cutting tip (up to 500 punches).
Caratteristiche e vantaggi
- Stainless steel cutting edges Stainless steel cutting edges (3.0 mm) enable manual coring, retrieval, and controlled ejection of the FTA™ punch.
- Designed to minimize contamination and carryover between samples when a cleaning procedure is followed.
Altre note
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)