Documenti fondamentali
WHA68761304
Filtri da siringa Whatman® GD/X
polysulfone membrane, pore size 0.45 μm, diam. 13 mm, non-sterile, pk of 50 ea
Sinonimo/i:
Whatman filter, Z286575, syringe filter, syringe filters
Scegli un formato
1 PKG
540,00 €
540,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 PKG
540,00 €
About This Item
Codice UNSPSC:
41104922
NACRES:
NB.24
540,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
polypropylene housing
polysulfone membrane
Sterilità
non-sterile
Caratteristiche
glass fiber prefilter
Confezionamento
pk of 50 ea
Produttore/marchio commerciale
Whatman 6876-1304
Whatman Article No. 28420516 (US reference)
Diametro
13 mm
Volume
≤10 mL
Dimensione pori
0.45 μm
Raccordi
female Luer inlet (lock)
male Luer outlet
Descrizione generale
Non-sterile syringe filters with a Polyethersulfone (PES) membrane, suitable for serum, plasma and tissue culture solutions. Whatman GD/X Syringe Filters are high-viscosity filters that incorporate a prefilter to enable filtration of viscous, dense or otherwise hard-to-filter material with high solids content as in environmental and food analysis samples. The prefilter contains a high-efficiency borosilicate glass filtration stack that will retain fine particles down to 0.7 μm, allowing difficult sample filtration with less hand force. It reduces the need for multiple syringe filters by processing three to seven times more sample volume than syringe filters without prefilters. The filters can be autoclaved.
Caratteristiche e vantaggi
  • Non sterile Whatman GD/X Syringe Filters combine a polypropylene housing, high-efficiency Whatman GMF 150 and GF/F pre-filter media, and one of the following membrane materials:
  • Polyethersulfone (PES) for serum, plasma and tissue culture
  • Polyvinylidene difluoride (PVDF) for HPLC
  • Cellulose acetate for biological solutions
  • Borosilicate glass microfiber
Compatibilità
Suitable for filtering environmental and food analysis samples. Suitable for particulate laden sample preparation.
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)